3-Amino-3-(3-chlorophenyl)propionic acid 98%

BLDPBD34613-250MGEA 7.3 EUR
BLDPBD34613-250MG BLDPBD34613-25G BLDPBD34613-5G BLDPBD34613-1G
3-Amino-3-(3-chlorophenyl)propionic acid 98%
3-Amino-3-(3-chlorophenyl)propionic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00463160
CAS-numero: 68208-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR