5-Amino-2-bromo-3-methylpyridine 95%

BLDPBD3585-25GEA 83.2 EUR
BLDPBD3585-25G BLDPBD3585-5G BLDPBD3585-1G BLDPBD3585-100G
5-Amino-2-bromo-3-methylpyridine 95%
5-Amino-2-bromo-3-methylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₆H₇BrN₂
MW: 187,04 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD03095085
CAS-numero: 38186-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR