3-Amino-4-fluorophenol 98%

BLDPBD40734-5GEA 26 EUR
BLDPBD40734-5G BLDPBD40734-25G BLDPBD40734-1G BLDPBD40734-10G BLDPBD40734-100G
3-Amino-4-fluorophenol 98%
3-Amino-4-fluorophenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₆H₆FNO
MW: 127,1200 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09033850
CAS-numero: 62257-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR