LXR-623 98%

BLDPBD408280-50MGEA 382 EUR
BLDPBD408280-50MG BLDPBD408280-10MG BLDPBD408280-25MG
LXR-623 98%
LXR-623

Storage condition: Sealed in dry, Room Temperature

Kaava: C₂₁H₁₂ClF₅N₂
MW: 422,78 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD16495812
CAS-numero: 875787-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR