3-Bromobenzylamine hydrochloride 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD41659-25GEA 87.3 EUR
BLDPBD41659-25G BLDPBD41659-100G BLDPBD41659-5G BLDPBD41659-1G
3-Bromobenzylamine hydrochloride 98%
3-Bromobenzylamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₉BrClN
MW: 222,52 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012856
CAS-numero: 39959-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR