N,N'-Diisopropylethylenediamine 98%

BLDPBD41662-25GEA 41.6 EUR
BLDPBD41662-25G BLDPBD41662-5G
N,N'-Diisopropylethylenediamine 98%
N,N'-Diisopropylethylenediamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₈H₂₀N₂
MW: 144,26 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008864
CAS-numero: 4013-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR