1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD42059-100GEA 14.6 EUR
BLDPBD42059-100G BLDPBD42059-500G BLDPBD42059-25G
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) 95%
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers)

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₈H₁₀O₃
MW: 154,17 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064863
CAS-numero: 85-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR