(3,5-Dichlorophenyl)methanesulphonyl chloride 97%
|
|
BLDPBD44915-250MGEA
53
EUR
BLDPBD44915-250MG
BLDPBD44915-1G
BLDPBD44915-100MG
(3,5-Dichlorophenyl)methanesulphonyl chloride 97%
(3,5-Dichlorophenyl)methanesulphonyl chloride
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₇H₅CI₃O₂S MW: 259,54 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD01631928 CAS-numero: 163295-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...