4-Chlorophenylglyoxal hydrate 97%

BLDPBD490926-1GEA 76.9 EUR
BLDPBD490926-1G BLDPBD490926-250MG
4-Chlorophenylglyoxal hydrate 97%
4-Chlorophenylglyoxal hydrate

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₈H₅ClO₂·H₂O
MW: 186,592 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08272327
CAS-numero: 859932-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR