2-Phenyl-9H-carbazole 97%

BLDPBD491294-100GEA 607 EUR
BLDPBD491294-100G BLDPBD491294-1G BLDPBD491294-25G BLDPBD491294-250MG BLDPBD491294-5G BLDPBD491294-0.025
2-Phenyl-9H-carbazole 97%
2-Phenyl-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₈H₁₃N
MW: 243,31 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD26402145
CAS-numero: 88590-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR