(S)-Mephenytoin 98%

BLDPBD49173-50MGEA 282 EUR
BLDPBD49173-50MG BLDPBD49173-250MG BLDPBD49173-25MG BLDPBD49173-100MG
(S)-Mephenytoin 98%
(S)-Mephenytoin
Kaava: C₁₂H₁₄N₂O₂
MW: 218,3 g/mol
CAS-numero: 70989-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR