4-Methoxy-3-nitrophenylboronic acid 98%

BLDPBD4919-250MGEA 41.6 EUR
BLDPBD4919-250MG BLDPBD4919-5G BLDPBD4919-100MG BLDPBD4919-1G
4-Methoxy-3-nitrophenylboronic acid 98%
4-Methoxy-3-nitrophenylboronic acid

Storage condition: Sealed in dry, 2-8°C

Kaava: C₇H₈BNO₅
MW: 196,9531 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD03092922
CAS-numero: 827614-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR