1-(2,6-Dimethylphenoxy)-2-propanone 95%

BLDPBD49786-250MGEA 42.6 EUR
BLDPBD49786-250MG BLDPBD49786-100MG BLDPBD49786-1G
1-(2,6-Dimethylphenoxy)-2-propanone 95%
1-(2,6-Dimethylphenoxy)-2-propanone

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₁H₁₄O₂
MW: 178,23 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00130266
CAS-numero: 53012-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR