Z-Thr(tBu)-OH 97%

BLDPBD517989-25GEA 127 EUR
BLDPBD517989-25G BLDPBD517989-250MG BLDPBD517989-100G BLDPBD517989-5G BLDPBD517989-10G BLDPBD517989-1G
Z-Thr(tBu)-OH 97%
Z-Thr(tBu)-OH

Storage condition: Sealed in dry, 2-8°C

Kaava: C₁₆H₂₃NO₅
MW: 309,4 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD06248909
CAS-numero: 16966-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR