1,1-Dimethylguanidinium sulphate 98%

BLDPBD52444-100GEA 76.9 EUR
BLDPBD52444-100G BLDPBD52444-1G BLDPBD52444-25G BLDPBD52444-5G
1,1-Dimethylguanidinium sulphate 98%
1,1-Dimethylguanidinium sulphate

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₆H₂₀N₆O₄S
MW: 272,33 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013131
CAS-numero: 598-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR