1,5-Dibromo-2,4-dimethoxybenzene 97%

BLDPBD53960-25GEA 308 EUR
BLDPBD53960-25G BLDPBD53960-250MG BLDPBD53960-5G BLDPBD53960-1G
1,5-Dibromo-2,4-dimethoxybenzene 97%
1,5-Dibromo-2,4-dimethoxybenzene

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₈Br₂O₂
MW: 295,956 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00061097
CAS-numero: 24988-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR