1,3-Diisopropenylbenzene 98% stabiloitu


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

BLDPBD54103-5GEA 6.3 EUR
BLDPBD54103-0.025G BLDPBD54103-5G BLDPBD54103-100G BLDPBD54103-25G
1,3-Diisopropenylbenzene 98% stabiloitu
1,3-Diisopropenylbenzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₁₂H₁₄
MW: 158,2396 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00008584
CAS-numero: 3748-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR