1,4-Dibromo-2,5-diiodobenzene 95%

BLDPBD54588-100GEA 657 EUR
BLDPBD54588-100G BLDPBD54588-25G BLDPBD54588-5G BLDPBD54588-1G
1,4-Dibromo-2,5-diiodobenzene 95%
1,4-Dibromo-2,5-diiodobenzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₆H₂Br₂I₂
MW: 487,6970 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00082898
CAS-numero: 63262-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR