1,4-Dikloori-9,10-antrakinoni 95%

BLDPBD54621-1GEA 216 EUR
BLDPBD54621-1G BLDPBD54621-250MG BLDPBD54621-5G
1,4-Dikloori-9,10-antrakinoni 95%
1,4-Dikloori-9,10-antrakinoni

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₄H₆Cl₂O₂
MW: 277,1022 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00192002
CAS-numero: 602-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR