1,4-Dimethoxy-2-butyne 98%

BLDPBD54699-100MGEA 18.7 EUR
BLDPBD54699-100MG BLDPBD54699-1G BLDPBD54699-250MG BLDPBD54699-5G
1,4-Dimethoxy-2-butyne 98%
1,4-Dimethoxy-2-butyne

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₆H₁₀O₂
MW: 114,14 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00048069
CAS-numero: 16356-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR