1-[(Ammoniooxy)methyl]-2,4-dichlorobenzene chloride 97%
|
|
BLDPBD55294-1GEA
18.7
EUR
BLDPBD55294-1G
BLDPBD55294-0.025G
BLDPBD55294-250MG
BLDPBD55294-5G
BLDPBD55294-25G
1-[(Ammoniooxy)methyl]-2,4-dichlorobenzene chloride 97%
1-[(Ammoniooxy)methyl]-2,4-dichlorobenzene chloride
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₇H₈Cl₃NO MW: 228,5000 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD01114586 CAS-numero: 51572-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...