2-(Hydroxymethyl)benzo[b]furan 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD56544-100MGEA 25 EUR
BLDPBD56544-100MG BLDPBD56544-250MG BLDPBD56544-1G BLDPBD56544-5G
2-(Hydroxymethyl)benzo[b]furan 95%
2-(Hydroxymethyl)benzo[b]furan

Storage condition: Sealed in dry, 2-8°C

Kaava: C₉H₈O₂
MW: 148,159 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00673995
CAS-numero: 55038-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR