1-Chloro-2-naphthol 97%

BLDPBD57766-10GEA 111 EUR
BLDPBD57766-10G BLDPBD57766-100G BLDPBD57766-5G BLDPBD57766-250MG BLDPBD57766-1G BLDPBD57766-25G
1-Chloro-2-naphthol 97%
1-Chloro-2-naphthol

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₇ClO
MW: 178,62 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00997084
CAS-numero: 633-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR