2,2'-Dibromoacetophenone 98%

BLDPBD5801-5GEA 76.9 EUR
BLDPBD5801-5G BLDPBD5801-1G BLDPBD5801-250MG
2,2'-Dibromoacetophenone 98%
2,2'-Dibromoacetophenone

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₈H₆Br₂O
MW: 277,9406 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00044660
CAS-numero: 49851-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR