2-Nitro-4-(trifluoromethyl)anisole 98%

BLDPBD5956-10GEA 10.4 EUR
BLDPBD5956-10G BLDPBD5956-5G BLDPBD5956-25G
2-Nitro-4-(trifluoromethyl)anisole 98%
2-Nitro-4-(trifluoromethyl)anisole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₆F₃NO₃
MW: 221,14 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007099
CAS-numero: 394-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR