1-Methyl-1H-indazole-5-boronic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD59613-25GEA 429 EUR
BLDPBD59613-25G BLDPBD59613-250MG BLDPBD59613-0.025G BLDPBD59613-5G BLDPBD59613-1G
1-Methyl-1H-indazole-5-boronic acid 98%
1-Methyl-1H-indazole-5-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₈H₉BN₂O₂
MW: 175,9800 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09870052
CAS-numero: 590418-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR