1-Phenacylpyridinium bromide 98%

BLDPBD60054-1GEA 9.4 EUR
BLDPBD60054-1G BLDPBD60054-25G BLDPBD60054-5G BLDPBD60054-100G
1-Phenacylpyridinium bromide 98%
1-Phenacylpyridinium bromide

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₃H₁₂BrNO
MW: 278,15 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00031995
CAS-numero: 16883-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR