Tetrahydrouridine 98%

BLDPBD601005-1MGEA 90.4 EUR
BLDPBD601005-1MG BLDPBD601005-10MG BLDPBD601005-5MG BLDPBD601005-25MG
Tetrahydrouridine 98%
Tetrahydrouridine
Kaava: C₉H₁₆N₂O₆
MW: 248,2331 g/mol
MDL numero: MFCD00063468
CAS-numero: 18771-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR