1-(tert-Butylamino)-3-chloropropan-2-ol hydrochloride 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD60491-1GEA 104 EUR
BLDPBD60491-1G BLDPBD60491-250MG
1-(tert-Butylamino)-3-chloropropan-2-ol hydrochloride 97%
1-(tert-Butylamino)-3-chloropropan-2-ol hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₁₆ClNO·ClH
MW: 202,122 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00191401
CAS-numero: 41052-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR