2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole 98+%

BLDPBD60592-250MGEA 64.5 EUR
BLDPBD60592-250MG BLDPBD60592-1G BLDPBD60592-100MG BLDPBD60592-5G
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole 98+%
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₇H₂₆Cl₂O₂
MW: 333,29 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD03093939
CAS-numero: 146370-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR