3-Chloro-(4-methoxycarbonyl)phenylboronic acid 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD6249-5GEA 65.5 EUR
BLDPBD6249-5G BLDPBD6249-25G BLDPBD6249-250MG BLDPBD6249-1G
3-Chloro-(4-methoxycarbonyl)phenylboronic acid 97%
3-Chloro-(4-methoxycarbonyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₈H₈BClO₄
MW: 214,4100 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD06659874
CAS-numero: 603122-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR