9,9'-Spirobi[9H-fluoren]-2-amine 98%

BLDPBD625745-1GEA 32.3 EUR
BLDPBD625745-1G BLDPBD625745-25G BLDPBD625745-5G
9,9'-Spirobi[9H-fluoren]-2-amine 98%
9,9'-Spirobi[9H-fluoren]-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₂₅H₁₇N
MW: 331,42 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD27956479
CAS-numero: 118951-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR