(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid 95%

BLDPBD62688-100MGEA 49.9 EUR
BLDPBD62688-100MG
(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid 95%
(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid

Storage condition: Sealed in dry, 2-8°C

Kaava: C₁₁H₉ClO₂S
MW: 240,706 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00052300
CAS-numero: 51527-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR