5-Bromo-2-(5-bromo-2-thienyl)pyridine 98%

BLDPBD627682-1GEA 97.7 EUR
BLDPBD627682-1G BLDPBD627682-50MG BLDPBD627682-100MG BLDPBD627682-5G BLDPBD627682-250MG
5-Bromo-2-(5-bromo-2-thienyl)pyridine 98%
5-Bromo-2-(5-bromo-2-thienyl)pyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₉H₅Br₂NS
MW: 319,01 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD29765747
CAS-numero: 136902-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR