5,6-Dibromo-2,1,3-benzothiadiazole 97%

BLDPBD629323-250MGEA 31.2 EUR
BLDPBD629323-250MG BLDPBD629323-1G BLDPBD629323-100MG BLDPBD629323-5G
5,6-Dibromo-2,1,3-benzothiadiazole 97%
5,6-Dibromo-2,1,3-benzothiadiazole
Kaava: C₆H₂Br₂N₂S
MW: 293,9700 g/mol
MDL numero: MFCD20257910
CAS-numero: 18392-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR