2-(Difluoromethoxy)benzylamine 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD63090-250MGEA 25 EUR
BLDPBD63090-250MG BLDPBD63090-1G
2-(Difluoromethoxy)benzylamine 98%
2-(Difluoromethoxy)benzylamine

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Kaava: C₈H₉F₂NO
MW: 173,16 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00236229
CAS-numero: 243863-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR