2-(Methylthio)benzimidazole 99%

BLDPBD63304-5GEA 39.5 EUR
BLDPBD63304-5G BLDPBD63304-10G BLDPBD63304-100G BLDPBD63304-25G
2-(Methylthio)benzimidazole 99%
2-(Methylthio)benzimidazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₈N₂S
MW: 164,2275 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005594
CAS-numero: 7152-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR