Methyl 3,5-Dihydroxy-4-methoxybenzoate 98%
|
|
BLDPBD633587-5GEA
420
EUR
BLDPBD633587-5G
BLDPBD633587-1G
BLDPBD633587-250MG
Methyl 3,5-Dihydroxy-4-methoxybenzoate 98%
Methyl 3,5-Dihydroxy-4-methoxybenzoate
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₉H₁₀O₅ MW: 198,17 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00017195 CAS-numero: 24093-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...