cis-6-Amino-cyclohex-3-enecarboxylic acid hydrochloride 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD633655-250MGEA 241 EUR
BLDPBD633655-250MG BLDPBD633655-100MG BLDPBD633655-1G
cis-6-Amino-cyclohex-3-enecarboxylic acid hydrochloride 97%
cis-6-Amino-cyclohex-3-enecarboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₁₁NO₂·ClH
MW: 177,629 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05863558
CAS-numero: 57266-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR