1H,1H,2H,2H-Nonafluorohexyl methacrylate 98% stabiloitu


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

BLDPBD63421-10GEA 28.1 EUR
BLDPBD63421-10G BLDPBD63421-5G BLDPBD63421-25G BLDPBD63421-1G BLDPBD63421-100G
1H,1H,2H,2H-Nonafluorohexyl methacrylate 98% stabiloitu
1H,1H,2H,2H-Nonafluorohexyl methacrylate

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₀H₉F₉O₂
MW: 332,17 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00236094
CAS-numero: 1799-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR