2-(9H-Carbazol-9-yl)phenylboronic acid 95%

BLDPBD635449-1GEA 55.1 EUR
BLDPBD635449-1G BLDPBD635449-10G BLDPBD635449-0.025 BLDPBD635449-25G BLDPBD635449-5G
2-(9H-Carbazol-9-yl)phenylboronic acid 95%
2-(9H-Carbazol-9-yl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₈H₁₄BNO₂
MW: 287,13 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD30543408
CAS-numero: 1189047-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR