(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate 97%

BLDPBD638603-1GEA 68.6 EUR
BLDPBD638603-1G BLDPBD638603-5G BLDPBD638603-250MG
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate 97%
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₇H₁₈F₃IO₃S
MW: 486,29 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD20264882
CAS-numero: 1204518-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR