4'-Cyano-4-dodecyloxybiphenyl 98%

BLDPBD640485-1GEA 56.2 EUR
BLDPBD640485-1G BLDPBD640485-5G BLDPBD640485-250MG
4'-Cyano-4-dodecyloxybiphenyl 98%
4'-Cyano-4-dodecyloxybiphenyl

Storage condition: Sealed in dry, Room Temperature

Kaava: C₂₅H₃₃NO
MW: 363,55 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01941019
CAS-numero: 57125-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR