2,2-Dimethyl-3-hexanone 99%
|
|
BLDPBD64520-25GEA
732
EUR
BLDPBD64520-25G
BLDPBD64520-5G
BLDPBD64520-1G
2,2-Dimethyl-3-hexanone 99%
2,2-Dimethyl-3-hexanone
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₈H₁₆O MW: 128,21 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00027140 CAS-numero: 5405-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...