2,3,4,5,6-Pentafluorostyrene 98% stabiloitu


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

BLDPBD64821-1GEA 8.4 EUR
BLDPBD64821-1G BLDPBD64821-5G BLDPBD64821-25G
2,3,4,5,6-Pentafluorostyrene 98% stabiloitu
2,3,4,5,6-Pentafluorostyrene

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₈H₃F₅
MW: 194,1 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00000300
CAS-numero: 653-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR