Coumarin 504T 98%

BLDPBD65211-1GEA 169 EUR
BLDPBD65211-1G
Coumarin 504T 98%
Coumarin 504T

Storage condition: Sealed in dry, Room Temperature

Kaava: C₂₂H₂₇NO₄
MW: 369,46 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00144013
CAS-numero: 113869-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR