L(+)-α-(4-Hydroxyphenyl)glycine 97%

BLDPBD6552-1GEA 6.3 EUR
BLDPBD6552-1G BLDPBD6552-100G BLDPBD6552-25G BLDPBD6552-5G BLDPBD6552-10G
L(+)-α-(4-Hydroxyphenyl)glycine 97%
L(+)-α-(4-Hydroxyphenyl)glycine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₈H₉NO₃
MW: 167,1600 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00065932
CAS-numero: 32462-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR