Methyl-4-aminosalicylate 97%

BLDPBD6554-500GEA 231 EUR
BLDPBD6554-500G BLDPBD6554-10G BLDPBD6554-25G BLDPBD6554-5G BLDPBD6554-100G
Methyl-4-aminosalicylate 97%
Methyl-4-aminosalicylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₈H₉NO₃
MW: 167,18 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00088091
CAS-numero: 4136-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR