2-((4-Methoxyphenyl)amino)-2-thioxoacetamide 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD6629-1GEA 89.4 EUR
BLDPBD6629-1G BLDPBD6629-250MG BLDPBD6629-5G
2-((4-Methoxyphenyl)amino)-2-thioxoacetamide 95%
2-((4-Methoxyphenyl)amino)-2-thioxoacetamide

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₁₀N₂O₂S
MW: 210,253 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00158777
CAS-numero: 71369-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR