2,4-Dibromobenzyl alcohol 97%
|
|
BLDPBD66593-100GEA
867
EUR
BLDPBD66593-100G
BLDPBD66593-250MG
BLDPBD66593-1G
BLDPBD66593-25G
BLDPBD66593-5G
2,4-Dibromobenzyl alcohol 97%
2,4-Dibromobenzyl alcohol
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₇H₆Br₂O MW: 265,98 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD09996906 CAS-numero: 666747-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...